Anti-LYVE1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-726 NB100-726SS
NOVUNB100-726EA 363 EUR
NOVUNB100-726 NOVUNB100-726SS
Anti-LYVE1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LYVE1 (lymphatic vessel endothelial hyaluronan receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR