Anti-Actin Mouse Monoclonal Antibody [clone: mAbGEa]
Proveedor: Novus Biologicals
NB100-74340 NB100-74340SS
NOVUNB100-74340EA 634 EUR
NOVUNB100-74340 NOVUNB100-74340SS
Anti-Actin Mouse Monoclonal Antibody [clone: mAbGEa]
Anticuerpos
The Actin Antibody (mAbGEa) from Novus Biologicals is a mouse monoclonal antibody to Actin. This antibody reacts with human, mouse, rat, bovine, chinese hamster, drosophila, fungi, plant, protozoa, rabbit, sheep, xenopus, yeast, zebrafish. The Actin Antibody (mAbGEa) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Simple Western ,
Type: Primary
Antigen: Actin
Clonality: Monoclonal
Clone: mAbGEa
Conjugation: Unconjugated
Epitope:
Host: Mouse
More Product Information
Order Now
ESPECIFICACIONES
Simbolo antígeno ACTA1 ACTA1
Environmentally Preferable
Nombre del antígeno Actin Alpha 1, Skeletal Muscle Actin Alpha 1, Skeletal Muscle
Sinónimos de antígenos ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1 ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado Unconjugated Unconjugated
Clon mAbGEa mAbGEa
Reactividad Human
Huésped Mouse Mouse
ID del gen 58 58
Isotipo IgM kappa IgM kappa
Western blot Yes Yes
ELISA Yes Yes
FluorescenciaInmunológica Yes Yes
Storage buffer PBS
Temperatura de almacenamiento Cool Cool
Shipping temperature Ice Ice