Anti-DGAT1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB110-41487 NB110-41487SS
NOVUNB110-41487EA 660 EUR
NOVUNB110-41487 NOVUNB110-41487SS
Anti-DGAT1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: DGAT1 (diacylglycerol O-acyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

ESPECIFICACIONES

Simbolo antígeno DGAT1
Environmentally Preferable
Nombre del antígeno Diacylglycerol O-acyltransferase 1
Sinónimos de antígenos ARGP1|Acyl coenzyme A|DIAR7|Diglyceride acyltransferase|ARAT|DGAT|EC 2.3.1.20|Diacylglycerol O-acyltransferase 1|ACT-related gene product 1
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
ID del gen 8694
Isotipo
Western blot Yes
FluorescenciaInmunológica Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR