Anti-TERF2 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB110-57130 NB110-57130SS
NOVUNB110-57130EA 700 EUR
NOVUNB110-57130 NOVUNB110-57130SS
Anti-TERF2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TERF2 (telomeric repeat binding factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR