Anti-LRRK2 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB110-58771 NB110-58771SS
NOVUNB110-58771EA 571 EUR
NOVUNB110-58771 NOVUNB110-58771SS
Anti-LRRK2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR