Anti-BECN2 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB110-60982 NB110-60982SS
NOVUNB110-60982EA 558 EUR
NOVUNB110-60982 NOVUNB110-60982SS
Anti-BECN2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: BECN2 (beclin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR