Anti-Napsin-A Mouse Monoclonal Antibody [clone: TMU-Ad02]
Proveedor: Novus Biologicals
NB110-68133 NB110-68133SS
NOVUNB110-68133EA 606 EUR
NOVUNB110-68133 NOVUNB110-68133SS
Anti-Napsin-A Mouse Monoclonal Antibody [clone: TMU-Ad02]
Anticuerpos
Type: Primary
Antigen: Napsin-A
Clonality: monoclonal
Clone: TMU-Ad02
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human