Anti-BECN1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB110-87318 NB110-87318SS
NOVUNB110-87318EA 662 EUR
NOVUNB110-87318 NOVUNB110-87318SS
Anti-BECN1 Rabbit Polyclonal Antibody
Anticuerpos
The Beclin 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Beclin 1. This antibody reacts with human, mouse, porcine, canine, primate. The Beclin 1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Simple Western

Type: Primary
Antigen: Beclin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Porcine, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR