Anti-NES Mouse Monoclonal Antibody [clone: 10C2]

Proveedor: Novus Biologicals
NB300-266 NB300-266SS
NOVUNB300-266EA 694 EUR
NOVUNB300-266 NOVUNB300-266SS
Anti-NES Mouse Monoclonal Antibody [clone: 10C2]
Anticuerpos
The Nestin Antibody (10C2) from Novus Biologicals is a mouse monoclonal antibody to Nestin. This antibody reacts with human, mouse (negative), rabbit (negative), rat (negative). The Nestin Antibody (10C2) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:200

Type: Primary
Antigen: Nestin
Clonality: Monoclonal
Clone: 10C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse (Negative), Rabbit (Negative), Rat (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR