Anti-Niemann-Pick C1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB400-148 NB400-148SS
NOVUNB400-148EA 655 EUR
NOVUNB400-148 NOVUNB400-148SS
Anti-Niemann-Pick C1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Niemann-Pick C1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR