Anti-NR1H4 Rabbit Polyclonal Antibody
Proveedor: Novus Biologicals
NB400-153 NB400-153SS
NOVUNB400-153EA 558 EUR
NOVUNB400-153 NOVUNB400-153SS
Anti-NR1H4 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NR1H4 (nuclear receptor subfamily 1, group H, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human