Anti-CALR Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB600-103 NB600-103SS
NOVUNB600-103EA 622 EUR
NOVUNB600-103 NOVUNB600-103SS
Anti-CALR Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CALR (calreticulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR