Anti-CD34 Rat Monoclonal Antibody [clone: MEC 14.7]

Proveedor: Novus Biologicals
NB600-1071 NB600-1071SS
NOVUNB600-1071EA 676 EUR
NOVUNB600-1071 NOVUNB600-1071SS
Anti-CD34 Rat Monoclonal Antibody [clone: MEC 14.7]
Anticuerpos
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: MEC 14.7
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR