Anti-PRKCA Mouse Monoclonal Antibody [clone: MC5]

Proveedor: Novus Biologicals
NB600-201 NB600-201SS
NOVUNB600-201EA 630 EUR
NOVUNB600-201 NOVUNB600-201SS
Anti-PRKCA Mouse Monoclonal Antibody [clone: MC5]
Anticuerpos
Type: Primary
Antigen: PRKCA (protein kinase C, alpha)
Clonality: monoclonal
Clone: MC5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR