Anti-FTCD Mouse Monoclonal Antibody [clone: 58K-9]

Proveedor: Novus Biologicals
NB600-412 NB600-412SS
NOVUNB600-412EA 612 EUR
NOVUNB600-412 NOVUNB600-412SS
Anti-FTCD Mouse Monoclonal Antibody [clone: 58K-9]
Anticuerpos
Type: Primary
Antigen: FTCD (formimidoyltransferase cyclodeaminase)
Clonality: monoclonal
Clone: 58K-9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR