Anti-HSPA5 Rabbit Polyclonal Antibody
Proveedor: Novus Biologicals
NBP1-06274 NBP1-06274SS
NOVUNBP1-06274EA 715 EUR
NOVUNBP1-06274 NOVUNBP1-06274SS
Anti-HSPA5 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now