Anti-OLLAS Epitope Tag Rat Monoclonal Antibody [clone: L2]
Proveedor: Novus Biologicals
NBP1-06713 NBP1-06713SS
NOVUNBP1-06713EA 735 EUR
NOVUNBP1-06713 NOVUNBP1-06713SS
Anti-OLLAS Epitope Tag Rat Monoclonal Antibody [clone: L2]
Anticuerpos
Type: Tag
Antigen: OLLAS Epitope Tag
Clonality: monoclonal
Clone: L2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Pan