Anti-B3GAT1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-19788 NBP1-19788SS
NOVUNBP1-19788EA 554 EUR
NOVUNBP1-19788 NOVUNBP1-19788SS
Anti-B3GAT1 Rabbit Polyclonal Antibody
Anticuerpos
The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta-1,3-Glucuronyltransferase 1 / B3GAT1. This antibody reacts with human, mouse, rat. The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: beta-1,3-Glucuronyltransferase 1/B3GAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR