Anti-SCD Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-26393 NBP1-26393SS
NOVUNBP1-26393EA 554 EUR
NOVUNBP1-26393 NOVUNBP1-26393SS
Anti-SCD Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SCD (stearoyl-CoA desaturase (delta-9-desaturase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR