Anti-HIF1A Mouse Monoclonal Antibody [clone: 162c]

Proveedor: Novus Biologicals
NBP1-30333 NBP1-30333SS
NOVUNBP1-30333EA 595 EUR
NOVUNBP1-30333 NOVUNBP1-30333SS
Anti-HIF1A Mouse Monoclonal Antibody [clone: 162c]
Anticuerpos
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: 162c
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR