Anti-USP9X Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-48321 NBP1-48321SS
NOVUNBP1-48321EA 582 EUR
NOVUNBP1-48321 NOVUNBP1-48321SS
Anti-USP9X Rabbit Polyclonal Antibody
Anticuerpos
The USP9x Antibody from Novus Biologicals is a rabbit polyclonal antibody to USP9x. This antibody reacts with human, mouse. The USP9x Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50

Type: Primary
Antigen: USP9x
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR