Anti-CCR2 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-48337SS NBP1-48337
NOVUNBP1-48337SSEA 275 EUR
NOVUNBP1-48337SS NOVUNBP1-48337
Anti-CCR2 Rabbit Polyclonal Antibody
Anticuerpos
The CCR2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCR2. This antibody reacts with human, rat. The CCR2 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: CCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno CCR2
Nombre del antígeno Chemokine (C-C motif) receptor 2
Environmentally Preferable
Sinónimos de antígenos CMKBR2|CKR2|MCP-1-R|CCR-2|CC-CKR-2|CKR2A|CCR2A|CD192|CCR2B|CKR2B
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
ID del gen 729230
Isotipo
Western blot Yes
FluorescenciaInmunológica Yes
Citometría de flujo Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR