Anti-NKX6-1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-49672 NBP1-49672SS
NOVUNBP1-49672EA 755 EUR
NOVUNBP1-49672 NOVUNBP1-49672SS
Anti-NKX6-1 Rabbit Polyclonal Antibody
Anticuerpos
The NKX6.1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NKX6.1. This antibody reacts with human, mouse. The NKX6.1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:100

Type: Primary
Antigen: NKX6.1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR