Anti-SIRT1 Mouse Monoclonal Antibody [clone: 1F3]

Proveedor: Novus Biologicals
NBP1-51641 NBP1-51641SS
NOVUNBP1-51641EA 636 EUR
NOVUNBP1-51641 NOVUNBP1-51641SS
Anti-SIRT1 Mouse Monoclonal Antibody [clone: 1F3]
Anticuerpos
The Sirtuin 1 / SIRT1 Antibody (1F3) from Novus Biologicals is a mouse monoclonal antibody to Sirtuin 1 / SIRT1. This antibody reacts with human, mouse, rat, primate. The Sirtuin 1 / SIRT1 Antibody (1F3) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000

Type: Primary
Antigen: SIRT1
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Primate
Order Now

ESPECIFICACIONES

Simbolo antígeno SIRT1 SIRT1
Nombre del antígeno Sirtuin 1 Sirtuin 1
Environmentally Preferable
Sinónimos de antígenos NAD-dependent Deacetylase Sirtuin-1|SIR2L1|SIRT1|SIR2-like Protein 1 SIR2L1|SIR2alpha|SIR2
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado Unconjugated Unconjugated
Clon 1F3 1F3
Reactividad Human
Huésped Mouse Mouse
ID del gen 23411 23411
Isotipo IgG1 IgG1
Western blot Yes Yes
ELISA Yes Yes
FluorescenciaInmunológica Yes Yes
Citometría de flujo Yes Yes
Temperatura de almacenamiento Cool Cool
Shipping temperature Ice Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR