Anti-CTNNB1 Mouse monoclonal antibody unconjugated [clone: 12F7]
Proveedor: Novus Biologicals
NBP1-54467 NBP1-54467SS
NOVUNBP1-54467EA 623 EUR
NOVUNBP1-54467 NOVUNBP1-54467SS
Anti-CTNNB1 Mouse monoclonal antibody unconjugated [clone: 12F7]
Anticuerpos
Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: monoclonal
Clone: 12F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now