Anti-EXOSC9 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-71702 NBP1-71702SS
NOVUNBP1-71702EA 554 EUR
NOVUNBP1-71702 NOVUNBP1-71702SS
Anti-EXOSC9 Rabbit Polyclonal Antibody
Anticuerpos
The Exosome Component 9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Exosome Component 9. This antibody reacts with human, mouse, chicken. The Exosome Component 9 Antibody has been validated for the following applications: Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:100

Type: Primary
Antigen: EXOSC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR