Anti-ABCB5 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-77689 NBP1-77689SS
NOVUNBP1-77689EA 554 EUR
NOVUNBP1-77689 NOVUNBP1-77689SS
Anti-ABCB5 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ABCB5 (ATP-binding cassette, sub-family B (MDR/TAP), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR