Anti-MMP9 Mouse Monoclonal Antibody [clone: 4A3]

Proveedor: Novus Biologicals
NBP2-13173 NBP2-13173SS
NOVUNBP2-13173EA 596 EUR
NOVUNBP2-13173 NOVUNBP2-13173SS
Anti-MMP9 Mouse Monoclonal Antibody [clone: 4A3]
Anticuerpos
Type: Primary
Antigen: MMP9 (matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase))
Clonality: monoclonal
Clone: 4A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR