Anti-NTRK3 Rabbit Polyclonal Antibody
Proveedor: Novus Biologicals
NBP2-15185 NBP2-15185SS
NOVUNBP2-15185EA 554 EUR
NOVUNBP2-15185 NOVUNBP2-15185SS
Anti-NTRK3 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NTRK3 (neurotrophic tyrosine kinase, receptor, type 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse