Anti-GFP Mouse monoclonal antibody unconjugated [clone: 4B10B2]

Proveedor: Novus Biologicals
NBP2-22111 NBP2-22111SS
NOVUNBP2-22111EA 580 EUR
NOVUNBP2-22111 NOVUNBP2-22111SS
Anti-GFP Mouse monoclonal antibody unconjugated [clone: 4B10B2]
Anticuerpos
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: monoclonal
Clone: 4B10B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR