Anti-MKI67 Mouse Monoclonal Antibody [clone: 8D5]

Proveedor: Novus Biologicals
NBP2-22112 NBP2-22112SS
NOVUNBP2-22112EA 644 EUR
NOVUNBP2-22112 NOVUNBP2-22112SS
Anti-MKI67 Mouse Monoclonal Antibody [clone: 8D5]
Anticuerpos
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: monoclonal
Clone: 8D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR