Anti-RTEL1 Rabbit Polyclonal Antibody
Proveedor: Novus Biologicals
NBP2-22360 NBP2-22360SS
NOVUNBP2-22360EA 623 EUR
NOVUNBP2-22360 NOVUNBP2-22360SS
Anti-RTEL1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: RTEL1 (regulator of telomere elongation helicase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse