Anti-TP53BP1 Mouse Monoclonal Antibody [clone: 6B3E10]

Proveedor: Novus Biologicals
NBP2-25028 NBP2-25028SS
NOVUNBP2-25028EA 642 EUR
NOVUNBP2-25028 NOVUNBP2-25028SS
Anti-TP53BP1 Mouse Monoclonal Antibody [clone: 6B3E10]
Anticuerpos
Type: Primary
Antigen: TP53BP1 (tumor protein p53 binding protein 1)
Clonality: Monoclonal
Clone: 6B3E10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR