Anti-Ascorbate Oxidase Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
200-4148S 200-4148-0100
ROCK200-4148SEA 226 EUR
ROCK200-4148S ROCK200-4148-0100
Anti-Ascorbate Oxidase Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Ascorbate Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Cucurbita
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR