Anti-IgG (H+L) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Rockland Immunochemicals
ROCK710-1202EA 277 EUR
ROCK710-1202
Anti-IgG (H+L) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR