Flat Sterlisation Pouches
Proveedor: SPS
91BOP01015 91BOP01634 91BOP02142 91BOP01220
SPSA91BOP01015EA 0 EUR
SPSA91BOP01015 SPSA91BOP01634 SPSA91BOP02142 SPSA91BOP01220
Flat Sterlisation Pouches
Bolsas Bolsas de esterilización
These pouches offer users an effective microbial barrier while guaranteeing the effective flow of the sterilizing agent and a peel-off system which provides aseptic conditions on opening. Through medical paper specially developed to meet the requirements of steam sterilization, these pouches have a 60 g/m² paper surface which is performance-equivalent in terms of resistance and barrier to 70 g/m² paper. Apart from the ecological aspect concerning the volume of waste to be treated, the reinforced 60 g/m² paper, combined with tear-proof 'shatterless' film, offers flexible packaging and easy opening that are incomparable for this type of packaging.
  • Medical grade 60 g/m² paper __plus__ tear-proof shatterless film
  • Double flow indicator (ISO 11140-1) intrafilm printed EO Steam and Gas
  • Wide range of sizes in all formats (flat, gusset and self-adhesive)
  • No curling on opening
  • Excellent peel-off opening
  • Peel-off direction printed on reels
Certificaciones: Conform to NF EN ISO 11607-1 and EN 868-5