Contact slides, BD Hycheck™
Proveedor: DIFCO BD
290001 290002
DIFC290001EA 45.4 EUR
DIFC290001 DIFC290002
Contact slides, BD Hycheck™
Medios Medios preparados
Hygiene contact slides used to assess the microbiological contamination of surfaces or fluids.
  • Double sided
  • Feature a hinged paddle that bends for easy sampling