BD BBL™ Prepared tubed and Mycoflask media
Proveedor: DIFCO BD
211887 211917 221715 220966 221039 221195 230910 211876 211886 212354 221812 215332 221788 215331 257176 221787 257107 220502 221823 297354
DIFC211887EA 240 EUR
DIFC211887 DIFC211917 DIFC221715 DIFC220966 DIFC221039 DIFC221195 DIFC230910 DIFC211876 DIFC211886 DIFC212354 DIFC221812 DIFC215332 DIFC221788 DIFC215331 DIFC257176 DIFC221787 DIFC257107 DIFC220502 DIFC221823 DIFC297354
BD BBL™ Prepared tubed and Mycoflask media
Medios Medios preparados
BBL™ Prepared Tubed Media are for the isolation and identification of microorganisms. Tubed media are ideal for cultures requiring prolonged incubation, since many can be incubated with caps tightly closed to prevent dehydration and subsequent inhibition of growth.
  • Reduce possible contamination
  • Special long-necked, rubber lined screw caps eliminate danger of contamination
  • Larger-sized tubes with wider diameters permit easy inoculation and handling of media