Anaerobic systems, Gas Pak
Proveedor: DIFCO BD
260411 260413 260628 270308 260457 260656 260662 271040 260626 260618 261181 271051 270303 260630 270124 260680 260683 260678 260001 260684 260672 260673 260674 260671 260685
DIFC260411EA 304.24 EUR
DIFC260411 DIFC260413 DIFC260628 DIFC270308 DIFC260457 DIFC260656 DIFC260662 DIFC271040 DIFC260626 DIFC260618 DIFC261181 DIFC271051 DIFC270303 DIFC260630 DIFC270124 DIFC260680 DIFC260683 DIFC260678 DIFC260001 DIFC260684 DIFC260672 DIFC260673 DIFC260674 DIFC260671 DIFC260685
Anaerobic systems, Gas Pak
Sistemas anaeróbicos
Designed for the generation of anearobic, microaerophilic (reduced oxygen), or CO₂-enriched environments. The BD GasPak™ 100 and 150 self-contained anaerobic systems consist of one polycarbonate jar (vented or non-vented), lid with O-ring gasket, catalyst reaction chamber, BBL room temperature catalyst charges, one rack, and one tube holder.
  • GasPak EZ Container Systems offer waterless, catalyst-free convenience for use in producing anaerobic, microaerophilic or C0₂-enriched environments
  • GasPak EZ Pouch Systems offer the convenience of pouches integrated into a complete kit with everything you need to generate a pouch based anaerobic, microaerophilic or C0₂-enriched environment
  • GasPak Pouch is a self-contained system which, after the addition of water, will produce an anaerobic or microaerophil atmosphere
The BD GasPak™ 100 system features two catalyst charges; the BD GasPak™ 150 system features ten charges. These systems use hydrogen and carbon dioxide generator envelopes and do not require a vacuum pump, gas tank, manometer, or pressure reducing valve. The BD GasPak™ 100 system jar holds up to twelve 100mm dishes. The BD GasPak™ 150 system jar holds up to twelve 150mm petri dishes or thirty-six 100mm dishes. The BD GasPak™ 150 vent kit consists of a threaded vent, rubber tube, and a tubing clamp for converting a non-vented system into a vented system.