Biochemical tests, DrySlide™
Proveedor: DIFCO BD
231746
231748
DIFC231746EA
417.75
EUR
DIFC231746
DIFC231748
Biochemical tests, DrySlide™
Tests para microbiología
Tests antígeno-anticuerpos
AK-AK, inmunoensayo
The BD BBL DrySlide™ provides the alternative to biochemical testing. Reagent is dried on the slide, eliminating the need for reagent addition.
- Each slide is divided into four large segments, allowing for multiple testing on one slide
- Indole, results in 30 seconds
- Oxidase, results in 20 seconds