The Lumitester™ PD-30 measures the hygiene status in seconds with on-site detection of microorganisms or organic residues. Extremely compact instrument which weighs only 235 g (battery and bag included). Swab and measure, that's all, so measuring time is only 8 seconds. Detects all organic-based contamination. Best suited for cleanliness control of manufacturing sites, also cleanliness control and hygiene training of kitchens. Defective cleaning is rapidly identified on site, thus helping to prevent incidents by prompting corrective re-cleaning procedures. This reliable monitoring of fluids, dry and moist surfaces increases sensitivity from patented measurement system for ATP and AMP. Measurement in RLU–Relative Light Units, data transfer via USB port on PC, graphical data evaluation and documentation of results. Display in 8 different languages, temperature compensation program for stable measurement at 10 to 40 °C, Definable threshold values to enable individual monitoring control points and the device can be returned for certified re-calibration for auditing purposes. The Excel-based software is made for comfortable international use and is simple to operate. 2000 storage spaces, 200 modes and auto-calibration and internal cleanliness check of the measurement chamber. CE marked and produced in accordance with ISO 9001.
LuciPac Pen / Lucipac Pen AQUA: Sealed, fracture-proof pads, which are applicable for fluids, dry and damp surfaces. The pads can be removed individually from closeable packaging (5×20 pads) and are highly stable at room temperature (shelf life of up to a year at temperatures of 2 to 8 °C). Patented simultaneous measurement of AMP and ATP brings optimal sensitivity. Detergent-tolerant enzymes- no inhibition through cleaning agent residue.
- Simple and safe to operate
- Patented simultaneous AMP and ATP measurement for increased sensitivity
- Detergent-tolerant luciferase for correct results
- The patented circulation technology increases the stability of the luminescence and extends the signal
- Fast and precise measurement method
Información de suministro: Lumitester™ PD-30 is supplied with transport bag, holder, 2× alkali batteries, cleaning brush, tape, short instructions plus CD-ROM with PC evaluation software.