Safety signs
Proveedor: Brady
814392 814446 814605 814273 814419 814604 816187 815710 815713 815551 815554 814915 814918 814265 814268 814756 814759 814597 814600 814438 814441 814411 814414 816031 814384 814387 816028
BRDY814392EA 14.6 EUR
BRDY814392 BRDY814446 BRDY814605 BRDY814273 BRDY814419 BRDY814604 116-0570 116-0572 116-0573 116-0574 116-0575 116-0576 116-0577 116-0578 116-0579 116-0580 116-0581 116-0560 116-0561 116-0562 116-0563 116-0564 116-0565 116-0569 116-0566 116-0567 116-0568
Safety signs
Señales
A comprehensive range of Emergency Escape and First Aid signs which help comply with current legislation.
  • Laminated polyester
  • Resistance to most chemical products and solvents
  • Resistance against extreme temperatures
  • Good weatherability and UV resistance
  • Excellent adhesion to low surface energy surfaces
  • Available in different materials and sizes