Screw caps
Proveedor: EP SCIENTIFIC
067-3800WBP 067-4800WBP 067-2410WBP
EPSC067-3800WBPEA 371 EUR
EPSC067-3800WBP EPSC067-4800WBP EPSC067-2410WBP
Screw caps
Tapas Tapones de botellas
LDPE-lined white caps.
  • For general-purpose HDPE bulk separate containers
  • Same high quality caps used on certified HDPE environmental sample containers
Información para pedidos: Bottles sold separately (IR013-series and IR012-series).