Anti-Ghrelin Mouse Monoclonal Antibody [clone: 5E611]

Proveedor: US Biological
G2033-74H-500 G2033-74H-250
USBIG2033-74H-500EA 612 EUR
USBIG2033-74H-500 USBIG2033-74H-250
Anti-Ghrelin Mouse Monoclonal Antibody [clone: 5E611]
Anticuerpos
Type: Primary
Antigen: Ghrelin
Clonality: Monoclonal
Clone: 5E611
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR