Anti-Glucagon Like Peptide 1 Mouse Monoclonal Antibody [clone: 6F118]

Proveedor: US Biological
G2040-13N-100 G2040-13N-25
USBIG2040-13N-100EA 654 EUR
USBIG2040-13N-100 USBIG2040-13N-25
Anti-Glucagon Like Peptide 1 Mouse Monoclonal Antibody [clone: 6F118]
Anticuerpos
Recommended dilution: ELISA: 1: 20000; Western Blot: 1:1000

Type: Primary
Antigen: Glucagon Like Peptide 1
Clonality: Monoclonal
Clone: 6F118
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mamalian,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR