Anti-CD103 Hamster Monoclonal Antibody [clone: 2E7]
Proveedor: US Biological
C2445-63R-200 C2445-63R-50
USBIC2445-63R-200EA 540 EUR
USBIC2445-63R-200 USBIC2445-63R-50
Anti-CD103 Hamster Monoclonal Antibody [clone: 2E7]
Anticuerpos
Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse,