Anti-Interferon Beta Rabbit Polyclonal Antibody
Proveedor: US Biological
140910 140910-100
USBI140910EA 485 EUR
USBI140910 USBI140910-100
Anti-Interferon Beta Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Interferon Beta
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine,