Anti-Zrp Mouse Monoclonal Antibody [clone: MN-7F2]

Proveedor: Cayman Chemical
100018-100 100018-500
CAYM100018-100EA 546 EUR
CAYM100018-100 CAYM100018-500
Anti-Zrp Mouse Monoclonal Antibody [clone: MN-7F2]
Anticuerpos
Type: Primary
Antigen: Zrp
Clonality: Monoclonal
Clone: MN-7F2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Atlantic Salmon, Rainbow Trout
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR