Anti-Vtg Mouse Monoclonal Antibody [clone: CK-1H11]

Proveedor: Cayman Chemical
100033-100 100033-500
CAYM100033-100EA 546 EUR
CAYM100033-100 CAYM100033-500
Anti-Vtg Mouse Monoclonal Antibody [clone: CK-1H11]
Anticuerpos
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: CK-1H11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Atlantic Cod, Carp, japanese medaka, Rainbow Trout, Tilapia
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR