Anti-Vtg Mouse Monoclonal Antibody [clone: ND-3C3]

Proveedor: Cayman Chemical
10007049-100 10007049-500
CAYM10007049-100EA 546 EUR
CAYM10007049-100 CAYM10007049-500
Anti-Vtg Mouse Monoclonal Antibody [clone: ND-3C3]
Anticuerpos
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: ND-3C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: american kestrel, barn swallow, common tern, domestic Chicken, japanese quail, Zebra finch
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR